Learn More
Macherey-Nagel™ Chromabond XTR Column
Shop All Macherey-Nagel Products
Click to view available options
Capacity (Metric):
150 mL
Unit Size:
Each
Specifications
Specifications
| Product Type | Chromabond XTR Column |
| Capacity (Metric) | 150 mL |
| Material | Polypropylene |
| Quantity | 10 Pack |
| Weight (Metric) | 37.5 g |