Learn More
Macherey-Nagel™ Chromabond™ BT 330 SiOH Column
Product Code. 15157065 Shop All Macherey-Nagel Products
Change view
Click to view available options
Length (Metric):
270 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15157065 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15157065 Supplier Macherey-Nagel™ Supplier No. 732967
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 02-10-2026
Specifications
Length (Metric) 270 mm
Type Flash Column
Diameter (Metric) Outer 60 mm
For Use With (Equipment) Biotage Isolera Systems