Learn More
Bryson Packaging™ Resealable Bags
Product Code. 10264594
Change view
Click to view available options
Width (Metric):
60 mm
140 mm
205 mm
280 mm
352 mm
Height (Metric):
60 mm
140 mm
280 mm
325 mm
405 mm
Product Type:
Sample Bag
Write-on Bag
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Product Type Unit Size
10264594 205 mm 280 mm Write-on Bag Pack of 100
10071424 140 mm 140 mm Sample Bag Pack of 100
11346934 205 mm 280 mm Sample Bag Pack of 100
10779005 352 mm 325 mm Sample Bag Pack of 100
11366934 280 mm 405 mm Sample Bag Pack of 100
10021664 60 mm 60 mm - Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10264594 Supplier Bryson Packaging™ Supplier No. BAJ340120J
  • ONLINE EXCLUSIVE PRICE
    33.60 EUR
    10.44 EUR / Pack of 100
    Save 23.16 EUR
    68% Off
Estimated Shipment: 11-12-2026
Transparent and excellent for shipping, storage and as sample bags. Bryson Packaging™ Resealable Bags are made from polyethylene.
  • Clear in color
  • Tight seal ensured
Specifications
Description Resealable Bag
Color Clear
For Use With (Application) Ideal for storing stationery and sending samples
Material Polyethylene
Quantity 100 Pack
Height (Metric) 280 mm
Product Type Write-on Bag
Width (Metric) 205 mm