Learn More
Choline hydroxide, 46% w/w aq. soln.
Description
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Choline hydroxide is used as a precursor molecule for the neurotransmitter acetylcholine, it is involved in several functions including muscle and memory muscle control. It was used as a basic catalyst for aldol condensation reactions.
Solubility
Fully miscible in water.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, air, acids.
Specifications
Specifications
| Density | 1.073 |
| Linear Formula | HOCH2CH2N(CH3)3OH |
| Refractive Index | 1.427 |
| Quantity | 100 mL |
| UN Number | UN3267 |
| Sensitivity | Air Sensitive |
| Solubility Information | Fully miscible in water. |
| IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;hydroxide |
| Formula Weight | 121.18 |
| Concentration or Composition (by Analyte or Components) | 46% w/w aq. soln. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.