missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cholic acid, sodium salt, 98+%

Product Code. p-3736023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10321061 100 g Glass bottle 100g
10667954 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10321061 Supplier Thermo Scientific Chemicals Supplier No. 405191000
Add to basket
This item is not returnable. View return policy

Cholic acid, sodium salt, CAS # 361-09-1, is a bile acid that can be non-denaturing detergent and can be used for membrane protein extraction. | CAS: 361-09-1 | C24H39NaO5 | 430.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description
• Cholic acid, sodium salt, is a bile acid that can aid digestion of fats and certain nutrients

Application
• It can facilitate digestion of lipids
• It is a non-denaturing detergent
• It can be used in combination with urea for extraction of membrane protein and protein complexes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361-09-1
Molecular Formula C24H39NaO5
Molecular Weight (g/mol) 430.56
MDL Number MFCD00150749,MFCD00064138
InChI Key NRHMKIHPTBHXPF-TUJRSCDTSA-M
Synonym sodium cholate, sodium 3,7,12-trihydroxycholan-24-oate, sodium cholate, reagent, ksc575s0b, 3,7,12-trihydroxycholan-24-oic acid sodium salt
PubChem CID 57349315
IUPAC Name sodium;(4R)-4-[(8R,9S,10S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
SMILES [Na+].C[C@H](CCC([O-])=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Color White
pH 7.5 to 9.5 (5% aq. soln.)
Physical Form Powder
Quantity 100 g
Assay Percent Range 99%
Merck Index 14, 2203
Formula Weight 430.55
Percent Purity 98.0 to 102.0% (on dried basis)
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (110°C)
Packaging Glass bottle
Chemical Name or Material Cholic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.