Learn More
Cholesteryl Erucate, TRC
Specifications
Specifications
| Chemical Name or Material | Cholesteryl Erucate |
| CAS | 24516-39-0 |
| Molecular Formula | C49H86O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C49H86O2/c1-8-10-12-14-16-18-20-22-27-40(28-23-21-19-17-15-13-11-9-2)47(50)51-42-33-35-48(6)41(37-42)29-30-43-45-32-31-44(39(5)26-24-25-38(3)4)49(45,7)36-34-46(43)48/h23,28-29,38-39,42-46H,1,8-22,24-27,30-37,40H2,2-7H3/b28-23-/t39-,42+,43+,44-,45+,46+,48+,49-/m1/s1 |
| SMILES | CCCCCCCC\C=C/[CH2](CCCCCCCCC[CH2])C(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C |
| IUPAC Name | [(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] (Z)-2-decyldodec-3-enoate |
| Molecular Weight (g/mol) | 707.21 |
| Formula Weight | 706.6628 |