missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol Glucuronide, TRC

Product Code. 30135215
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135215 10 mg 10mg
30134287 25 mg 25mg
30006307 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135215 Supplier Toronto Research Chemicals Supplier No. TRCC17435010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17435-78-8
Molecular Formula C33 H54 O7
Molecular Weight (g/mol) 562.778
IUPAC Name (2S,3S,4S,5R,6R)-6-[[(3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C33H54O7/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(13-15-32(20,4)25(22)14-16-33(23,24)5)39-31-28(36)26(34)27(35)29(40-31)30(37)38/h9,18-19,21-29,31,34-36H,6-8,10-17H2,1-5H3,(H,37,38)/t19-,21+,22+,23-,24+,25+,26+,27+,28-,29+,31-,32+,33-/m1/s1
Formula Weight 562.387
Chemical Name or Material Cholesterol Glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.