Learn More
Chlorzoxazone USP Related Compound A, Mikromol™
Description
Chlorzoxazone USP Related Compound A
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4-chlorophenol |
| CAS | 95-85-2 |
| Analyte or Component Names | 2-Amino-4-chlorophenol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H6 Cl N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-Amino-4-chlorophenol, 1-Amino-2-hydroxy-5-chlorobenzene, 1-Hydroxy-2-amino-4-chlorobenzene, 2-Hydroxy-5-chloroaniline, 4-Chloro-2-aminophenol, 4-Chloro-o-aminophenol, 5-Chloro-2-hydroxyaniline, C.I. 76525, C.I. Oxidation Base 18, Fouramine PY, NSC 247814, o-Amino-p-chlorophenol, p-Chloro-o-aminophenol, Chlorzoxazone Related Compound A |
| InChI Formula | InChI=1S/C6H6ClNO/c7-4-1-2-6(9)5(8)3-4/h1-3,9H,8H2 |
| SMILES | Nc1cc(Cl)ccc1O |
| IUPAC Name | 2-amino-4-chlorophenol |
| Show More |