missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine hydrochloride, 100.9%, MP Biomedicals™

Product Code. 11455931 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455931 5 g 5g
11465931 25 g 25g
11475931 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455931 Supplier MP Biomedicals Supplier No. 0219032605
  • ONLINE EXCLUSIVE PRICE
    129.00 EUR
    122.85 EUR / 5g
    Save 6.15 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 69-09-0
Molecular Formula C17H20Cl2N2S
Molecular Weight (g/mol) 355.321
InChI Key FBSMERQALIEGJT-UHFFFAOYSA-N
Synonym 2-Chloro-10-[3-dimethylamino-propyl] phenothiazine, 2-Chloro-N, N- dimethyl-10H- phenothiazine-10- propanamine hydrochloride
PubChem CID 6240
ChEBI CHEBI:3649
IUPAC Name 3-(2-chlorophenothiazin-10-yl)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCN1C2=CC=CC=C2SC3=C1C=C(C=C3)Cl.Cl

Specifications

Assay Percent Range 1.009
Physical Form Powder/Solution
Color Colorless or White
Quantity 5 g
Formula Weight 355.3
Chemical Name or Material Chlorpromazine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.