Learn More
Chlorotrimethylsilane, Sigma-Aldrich™
Product Code. 15814451
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814451 500 mL 500mL
15804451 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814451 missing translation for 'mfr' Sigma Aldrich missing translation for 'supplierNo' 8187370500
 Restricted Item
Estimated Shipment: 23-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-77-4
Specifications
Melting Point -58°C
Density 0.86 g/cm3 at 20°C
Boiling Point 57°C/1013 hPa
Molecular Formula (CH3)3SiCl
MDL Number MFCD00000502
Quantity 500 mL
Synonym Chlorotrimethylsilane, Trimethylchlorosilane, Trimethylsilyl chloride, TMCS, Chlorotrimethylsilane
Molecular Weight (g/mol) 108.64
Grade Research
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CHLOROTRIMETHYLSILANE POUR LA SYNTHESE
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H331-Toxic if swallowed or if inhaled
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.