missing translation for 'onlineSavingsMsg'
Learn More

Chlorophyllin Copper Trisodium Salt (Technical Grade), TRC

Product Code. 30011124
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011124 25 g 25g
30009777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011124 Supplier Toronto Research Chemicals Supplier No. TRCC38023525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11006-34-1
Formula Weight 723.1233
InChI Formula InChI=1S/C34H36N4O6.Cu.3Na/c1-7-19-15(3)23-12-25-17(5)21(9-10-29(39)40)32(37-25)22(11-30(41)42)33-31(34(43)44)18(6)26(38-33)14-28-20(8-2)16(4)24(36-28)13-27(19)35-23;;;;/h7,12-14,17,21H,1,8-11H2,2-6H3,(H5,35,36,37,38,39,40,41,42,43,44);;;;/q;+2;3*+1/p-5
IUPAC Name copper;trisodium;3-[20-(carboxylatomethyl)-18-(dioxidomethylidene)-8-ethenyl-13-ethyl-3,7,12,17-tetramethyl-2,3-dihydroporphyrin-23-id-2-yl]propanoate
Molecular Formula C34 H31 Cu N4 O6 . 3 Na
Molecular Weight (g/mol) 724.15
SMILES [Na+].[Na+].[Na+].CCC1=C(C)C2=CC3=C(C=C)C(=C4C=C5C(C)C(CCC(=O)[O-])C6=N5[Cu+2]7(N2=C1C=C8C(=C(C(=O)[O-])C(=C6CC(=O)[O-])[N-]78)C)[N-]34)C
Synonym Cuprate(3-), [(7S,8S)-3-carboxy-5-(carboxymethyl)-13-ethenyl-18-ethyl-7,8-dihydro-2,8,12,17-tetramethyl-21H,23H-porphine-7-propanoato(5-)-κN21,κN22,κN23,κN24]-, sodium (1:3), (SP-4-2)-, Chlorine e6, copper deriv., trisodium salt (6CI), Chlorophyllin, copper sodium complex (8CI), Cuprate(3-), [(7S,8S)-3-carboxy-5-(carboxymethyl)-13-ethenyl-18-ethyl-7,8-dihydro-2,8,12,17-tetramethyl-21H,23H-porphine-7-propanoato(5-)-κN21,κN22,κN23,κN24]-, trisodium, (SP-4-2)- (9CI), JPM 01-03B, Rovisome I, Sodium-copper chlorin e6
Chemical Name or Material Chlorophyllin copper trisodium

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.