CAS RN 11006-34-1
CAS RN 11006-34-1
IUPAC Name:
chlorophyllin
Synonyms:
chlorophyllin
Molecular Weight (g/mol):
724.16
Molecular Formula:
C34H31CuN4Na3O6
InChi Key:
HWDGVJUIHRPKFR-UHFFFAOYNA-I
SMILES:
[Na+].[Na+].[Na+].[Cu++].CCC1=C(C)C2=N/C/1=C\C1=C(C)C(=C([O-])[O-])C(=N1)\C(CC([O-])=O)=C1/N=C(\C=C3/[N-]/C(=C\2)C(C=C)=C3C)C(C)C1CCC([O-])=O
1
–
3
of
3
results
1
Chlorophyllin, coppered trisodium salt
CAS: 11006-34-1 Molecular Formula: C34H31CuN4Na3O6 MDL Number: MFCD00012149
3
Chlorophyllin sodium copper salt, MedChemExpress
MedChemExpress Sodium copper chlorophyllin A is a semi-synthetic mixture of sodium copper salts derived from chlorophyll, primarily used as a food additive and in alternative medicine.