missing translation for 'onlineSavingsMsg'
Learn More

Chloroneb, TRC

Product Code. 30359251
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359251 100 mg 100mg
30355657 1 g 1g
30349428 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30359251 Supplier Toronto Research Chemicals Supplier No. TRCC370935100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2675-77-6
Molecular Formula C8 H8 Cl2 O2
Molecular Weight (g/mol) 207.05
Synonym 1,4-Dichloro-2,5-dimethoxybenzene, 2,5-Dichloro-1,4-dimethoxybenzene, Chloroneb, Demosan, Demosan 65W, FloPro D, NSC 151546, Soil Fungicide 1823
IUPAC Name 1,4-dichloro-2,5-dimethoxybenzene
SMILES COc1cc(Cl)c(OC)cc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8Cl2O2/c1-11-7-3-6(10)8(12-2)4-5(7)9/h3-4H,1-2H3
Formula Weight 205.9901
Chemical Name or Material Chloroneb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.