Learn More
(Chloromethyl)diphenylphosphine Oxide, TRC
Product Code. 30365263
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30365263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30365263 Supplier Toronto Research Chemicals Supplier No. TRCC369210100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1806-49-1
Molecular Formula C13H12ClOP
Quantity 100 mg
Synonym (Chloromethyl)diphenyl-phosphine Oxide
InChI Formula InChI=1S/C13H12ClOP/c14-11-16(15,12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10H,11H2
SMILES ClCP(=O)(c1ccccc1)c2ccccc2
IUPAC Name [chloromethyl(phenyl)phosphoryl]benzene
Molecular Weight (g/mol) 250.66
Formula Weight 250.0314
Chemical Name or Material (Chloromethyl)diphenylphosphine Oxide