missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Supelco™

Product Code. 32237611
Change view
Click to view available options
Quantity:
1 L
Unit Size:
2.50 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237611 1 L 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237611 Supplier Supelco Supplier No. 1024321000
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For gas chromatography ECD and FID SupraSolv™

SupraSolv™ ECD and FID is a high-purity solvent grade specifically formulated for sample preparation in gas chromatography with electron capture (ECD) and flame ionization (FID) detection.ECD and FID impose distinct and stringent solvent purity requirements. The electron capture detector is exquisitely sensitive to electronegative species - particularly halogenated compounds - at sub-nanogram concentrations, while the flame ionization detector responds to any carbon-containing organic impurity undergoing combustion. SupraSolv™ ECD and FID solvents are therefore subjected to rigorous purification and detector-specific testing protocols to ensure that intrinsic solvent impurities remain below the respective detection thresholds of both detector types simultaneously.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chloroform
Melting Point -63°C (lit.)
CAS 67-66-3
Density 1.492 g/mL at 25°C (lit.)
Quantity 1 L
Synonym TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane, Methylidyne trichloride, Trichloromethane
Boiling Point 60.5°C to 61.5°C (lit.)
Molecular Weight (g/mol) 119.38
Grade Not Available
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.