Learn More
Chloroform, Supelco™
Product Code. 15837110
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15837110 2.5 L 2.5 L
15827110 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15837110 Supplier Supelco Supplier No. 1024422500
 Restricted Item
Estimated Shipment: 19-11-2026
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
For analysis (for determinations with dithizone)
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-66-3
Specifications
Melting Point −63°C (lit.)
Density 1.492 g/mL at 25°C
Quantity 2.5 L
MDL Number MFCD00000826
Synonym TCM, Trichloromethane, Methane trichloride, Methyl trichloride, Methylidyne trichloride, Trichloromethane, Methylidyne trichloride, Trichloromethane
Boiling Point 60.5°C to 61.5°C
Molecular Weight (g/mol) 119.38
Grade Research
Chemical Name or Material Chloroform