Learn More
Chlorocyclohexane, Sigma-Aldrich™
Product Code. 15894511
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894511 1 L 1 L
15884511 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15894511 Supplier Sigma Aldrich Supplier No. 8202871000
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 542-18-7
Spezifikation
Melting Point -44°C
Density 1.00 g/cm3 at 20°C
Boiling Point 141°C to 143°C/1013 hPa
MDL Number MFCD00003822
Quantity 1 L
Synonym Chlorocyclohexane, Cyclohexyl chloride, Chlorocyclohexane
Molecular Weight (g/mol) 118.6
Grade Research