Learn More
Chlorobutanol Hemihydrate, Mikromol™
Product Code. 17953366
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17953366 Supplier Mikromol Supplier No. MM0535.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Chlorobutanol Hemihydrate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Acetonchloroform
CAS 6001-64-5
Analyte or Component Names Acetonchloroform
Shipping Condition Room Temperature
Molecular Formula 2 C4 H7 Cl3 O . H2 O
Certifications/Compliance ISO 17025
Synonym 2-Propanol, 1,1,1-trichloro-2-methyl-, hemihydrate (8CI), 1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, Chlorobutanol hemihydrate
InChI Formula InChI=1S/2C4H7Cl3O.H2O/c2*1-3(2,8)4(5,6)7;/h2*8H,1-2H3;1H2
SMILES O.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
IUPAC Name 1,1,1-trichloro-2-methylpropan-2-ol;hydrate
Molecular Weight (g/mol) 372.93
Quantity 500 mg
Formula Weight 369.9231 g/mol