missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, Sigma-Aldrich™

Product Code. 15838650
Change view
Click to view available options
Quantity:
100 mL
1 L
25 L
Unit Size:
1 L
100mL
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838650 25 L 25 L
15818650 100 mL 100mL
15828650 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15838650 Supplier Sigma Aldrich Supplier No. 8017919025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Chlorobenzeneis a colorless and flammable aromatic compound. In various industries,chlorobenzene is primarily used as a solvent, a degreasing agent, and as achemical intermediate.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7

Specifications

Melting Point -45°C
Density 1.11 g/cm3 at 20°C
Quantity 25 L
MDL Number MFCD00000530
Synonym Chlorobenzene, Phenyl chloride, Chlorobenzene
Boiling Point 132°C/1013 hPa
Molecular Weight (g/mol) 112.56
Grade Research
Chemical Name or Material Chlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.