missing translation for 'onlineSavingsMsg'
Learn More

Chlormethiazole Hydrochloride, TRC

Product Code. 30134926
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134926 100 mg 100mg
17910365 5 mg 5mg
17910495 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134926 Supplier Toronto Research Chemicals Supplier No. TRCC602013100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6001-74-7
Molecular Formula C6 H8 Cl N S . Cl H
Molecular Weight (g/mol) 198.11
Synonym Thiazole, 5-(2-chloroethyl)-4-methyl-, hydrochloride (1:1), Thiazole, 5-(2-chloroethyl)-4-methyl-, hydrochloride (6CI,7CI,8CI,9CI), 5-(2-Chloroethyl)-4-methylthiazole hydrochloride, Chlormethiazole hydrochloride, Clomethiazole hydrochloride
IUPAC Name 5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
SMILES Cl.Cc1ncsc1CCCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C6H8ClNS.ClH/c1-5-6(2-3-7)9-4-8-5;/h4H,2-3H2,1H3;1H
Formula Weight 196.9833
Chemical Name or Material Clomethiazole Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.