missing translation for 'onlineSavingsMsg'
Learn More

Chlormethiazole hydrochloride, MedChemExpress

Product Code. 16484197
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484197 50 mg 50mg
16478318 10 mg 10mg
16478046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16484197 Supplier Medchem Express Supplier No. HYA0296
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Clomethiazole hydrochloride is a anticonvulsant. Clomethiazole hydrochloride is neuroprotective and prevents the degeneration of serotonergic nerve terminals induced by 3,4-methylenedioxymethamphetamine (MDMA).

Clomethiazole hydrochloride is a anticonvulsant. Clomethiazole hydrochloride is neuroprotective and prevents the degeneration of serotonergic nerve terminals induced by 3,4-methylenedioxymethamphetamine (MDMA).

Specifications

Chemical Name or Material Chlormethiazole hydrochloride
Physical Form Solid
CAS 6001-74-7
Color Brownish Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H9Cl2NS
For Use With (Application) Neuroscience-Neuromodulation
Synonym Clomethiazole hydrochloride
Solubility Information DMSO : 125 mg/mL (630.96 mM; Need ultrasonic) ∣H2O : 100 mg/mL (504.77 mM; Need ultrasonic)
SMILES CC1=C(CCCl)SC=N1.Cl
Molecular Weight (g/mol) 198.11
Formula Weight 198.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.