Learn More
Sherwood Scientific™ Chloride Standard Solution, 200mg/L
Product Code. 12457570
Change view
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
600mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12457570 6 x 100 mL 600mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12457570 Supplier Sherwood Scientific™ Supplier No. 00156203
 Restricted Item
Estimated Shipment: 02-12-2026
Formulated for use with the Model 926 Chloride Analyzer. Sherwood Scientific Chloride Standard Solution, 200mg/L is ideal for determination of chloride ions using coulometric titration. Comes in a set of six 100ml vials.
The measurement of salt (Sodium Chloride) concentration in the Food and Dairy Industry is universal. Salt and salinity also represents serious contamination in industrial processes. The sensitivity of the coulometric method enables measurement in the ppm range in boiler feed water, polymer washes, bore hole slurries, as well as soils analysis and others.
The Model 926 method is based on a coulometric titration and is an absolute method where the reagent, the silver ions, is precisely and quantitatively generated at the time of the analysis by passing a constant current between donor electrodes. The end point is detected by the use of sensing electrodes which measure the change in solution conductivity which occurs when excess silver ions are present in solution.
Specifications
Color Clear, Colorless
Concentration 200mg/L
Type Chloride Standard Solution, 200mg/L
Packaging Vials
Quantity 6 x 100 mL