Learn More
Sherwood Scientific™ Chloride Analyzer M926 III
Product Code. 12687087
Change view
Click to view available options
For Use With (Application):
Measures Active Salt
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
12687087 Measures Active Salt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12687087 Supplier Sherwood Scientific™ Supplier No. 92600009
  • ONLINE EXCLUSIVE PRICE
    6860.00 EUR
    6322.88 EUR / Pack of 1
    Save 537.12 EUR
    7% Off
Estimated Shipment: 29-12-2026
Data manipulation with active salt
The models 926 and 926S are equipped with RS232 serial and USB outputs which enable them to be linked to a suitable printer or to be interfaced to a PC hosting Active Salt software, developed to enhance the model 926 chloride analyser. Designed initially for the cheese industry, the software may be applied to any food product requiring salt analysis and documentation. The software may be configured by the operator to suit their particular laboratory requirements. The active salt package can collect data directly from a balance with RS232 output together with the chloride analyser results. Some of the advantages are:
  • No transcription errors
  • No time wasted doing manual calculations
  • No need to achieve specific sample and diluent weights
  • Automatic data storage and analysis report generation
  • Chloride meters use the colorimetric titration method where silver ions are generated in-situ and react with chloride ions
  • End point precisely detected
  • No reagent needing recalibration
  • Economical per test
  • Connects to printer/PC
  • Supplied with printer cable RS232, acid buffer, three anodes, three electrodes, two marked beakers, electrode cleaning polish, stirrer and operating manual Mark III 926: for industrial applications with mg/L units. Mark III 926S: for clinical applications withmmol/L units. Suitable for salt in food, sweat test (reference method), waste water and biological fluids.
Specifications
Type Chloride Analyzer
For Use With (Application) Measures Active Salt