missing translation for 'onlineSavingsMsg'
Learn More

Chlorhexidine Free Base, >85%, MP Biomedicals™

Product Code. 11420432 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420432 100 g 100g
11400432 5 g 5g
11410432 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420432 Supplier MP Biomedicals Supplier No. 0219135880
  • ONLINE EXCLUSIVE PRICE
    7660.00 EUR
    7293.30 EUR / 100g
    Save 366.70 EUR
    4% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Molecular biology; Cell biology

Chemical Identifiers

CAS 55-56-1
Molecular Formula C22H30Cl2N10
Molecular Weight (g/mol) 505.452
MDL Number MFCD00009673
InChI Key GHXZTYHSJHQHIJ-UHFFFAOYSA-N
Synonym chlorhexidine, chlorhexidinum, chlorhexidin, nolvasan, rotersept, tubulicid, fimeil, hexadol, soretol, cloresidina
PubChem CID 9552079
ChEBI CHEBI:3614
IUPAC Name (1E)-2-[6-[[amino-[(E)-[amino-(4-chloroanilino)methylidene]amino]methylidene]amino]hexyl]-1-[amino-(4-chloroanilino)methylidene]guanidine
SMILES C1=CC(=CC=C1NC(=NC(=NCCCCCCN=C(N)N=C(N)NC2=CC=C(C=C2)Cl)N)N)Cl

Specifications

Color White
Quantity 100 g
Physical Form Solid
Assay Percent Range >85%
Formula Weight 505.5
Chemical Name or Material Chlorhexidine Free Base, Free Base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.