Learn More
Chlorantraniliprole, TRC
Product Code. 30134916
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134916 500 mg 500mg
30132897 1 g 1g
17962695 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134916 Supplier Toronto Research Chemicals Supplier No. TRCC325380500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 500008-45-7
Molecular Formula C18 H14 Br Cl2 N5 O2
Molecular Weight (g/mol) 483.15
Synonym 3-Bromo-N-[4-chloro-2-methyl-6-[(methylamino)carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-1H-pyrazole-5-carboxamide, Acelepryn, Altacor, Altacor 35WG, Altriset, Calteryx, Chlorantraniliprole, Coragen, Coragen 20CS, DKI 0001, DPX-E 2Y45, Dermacor X 100, E 2Y45, Ferterra, Prevathon, Rynaxypyr
IUPAC Name 5-bromo-N-[4-chloro-2-methyl-6-(methylcarbamoyl)phenyl]-2-(3-chloropyridin-2-yl)pyrazole-3-carboxamide
SMILES CNC(=O)c1cc(Cl)cc(C)c1NC(=O)c2cc(Br)nn2c3ncccc3Cl
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C18H14BrCl2N5O2/c1-9-6-10(20)7-11(17(27)22-2)15(9)24-18(28)13-8-14(19)25-26(13)16-12(21)4-3-5-23-16/h3-8H,1-2H3,(H,22,27)(H,24,28)
Formula Weight 480.9708
Percent Purity >95%
Chemical Name or Material Chlorantraniliprole
Grade HPLC