missing translation for 'onlineSavingsMsg'
Learn More

Chlorantraniliprole, MedChemExpress

Product Code. 16494708
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494708 10 mg 10mg
16475536 100 mg 100mg
16401007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494708 Supplier Medchem Express Supplier No. HY112820
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Chlorantraniliprole is an insecticide that potently and selectively activates insect ryanodine receptor, with EC50s of 40 nM and 50 nM for Drosophila melanogaster and H. virescens ryanodine receptor, and ∼300-fold more potent than that in the mouse myoblast cell line, C2C12 (EC50, 14 μM).

Chlorantraniliprole is an insecticide that potently and selectively activates insect ryanodine receptor, with EC50s of 40 nM and 50 nM for Drosophila melanogaster and H. virescens ryanodine receptor, and ∼300-fold more potent than that in the mouse myoblast cell line, C2C12 (EC50, 14 μM).

Specifications

Chemical Name or Material Chlorantraniliprole
CAS 500008-45-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H14BrCl2N5O2
Quantity 10 mg
Solubility Information DMSO : ≥ 62.5 mg/mL (129.36 mM)
SMILES O=C(C1=CC(Br)=NN1C2=NC=CC=C2Cl)NC3=C(C(NC)=O)C=C(Cl)C=C3C
Molecular Weight (g/mol) 483.15
Formula Weight 483.15
Percent Purity 95.0%
Grade Research
Physical Form Powder
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.