missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, 99%, MP Biomedicals™

Product Code. 11462111 Shop All MP Biomedicals Products
Click to view available options
Quantity:
20 mg
Unit Size:
20mg
This item is not returnable. View return policy

Product Code. 11462111

Brand: MP Biomedicals 0219478720

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 56-75-7
Color White to Cream
Density 1.586g/ml
Molecular Formula C11H12Cl2N2O5
Quantity 20 mg
Synonym chloramphenicol, chlornitromycin, chloromycetin, levomycetin, chlorocid, halomycetin, levomicetina, alficetyn, fenicol, globenicol
InChI Key WIIZWVCIJKGZOK-RKDXNWHRSA-N
SMILES C1=CC(=CC=C1C(C(CO)NC(=O)C(Cl)Cl)O)[N+](=O)[O-]
IUPAC Name 2,2-dichloro-N-[(1R,2R)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-yl]acetamide
Molecular Weight (g/mol) 323.126
PubChem CID 5959
ChEBI CHEBI:17698
Formula Weight 323.13
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Chloramphenicol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.