missing translation for 'onlineSavingsMsg'
Learn More

Chloramine T Sodium Salt Trihydrate Crystalline MP Biomedicals

Product Code. 11466512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466512 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11466512 Supplier MP Biomedicals Supplier No. 0215011583
  • ONLINE EXCLUSIVE PRICE
    89.30 EUR
    85.05 EUR / 250g
    Save 4.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Type Chloramine T Sodium Salt
Physical Form Crystalline
CAS 127-65-1
Quantity 250 g
Molecular Formula C7H7ClNNaO2S
Synonym N-Chloro-p-toluenesulfonamide
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
Molecular Weight (g/mol) 227.638
PubChem CID 3641960
ChEBI CHEBI:53767
Formula Weight 281.7
Chemical Name or Material Chloramine T Sodium Salt, Trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.