missing translation for 'onlineSavingsMsg'
Learn More

Chloramine T sodium salt trihydrate, 100.29%, MP Biomedicals™

Product Code. p-4404627 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476512 500 g 500g
11486512 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476512 Supplier MP Biomedicals Supplier No. 0215011590
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-65-1
Molecular Formula C7H7ClNNaO2S
Molecular Weight (g/mol) 227.638
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
Synonym chloramine-t, chloramine t, chloralone, chlorasan, chlorozone, acti-chlore, chloraseptine, chlorazene, chlorazone, chlorseptol
PubChem CID 3641960
ChEBI CHEBI:53767
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]

Specifications

Color White
Density Bulk density 540 to 680kg/m3
Assay Percent Range 1.0029
Quantity 500 g
Formula Weight 281.7
Physical Form Fine Powder
Chemical Name or Material N-Chloro-p-toluenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.