Learn More
Chloramine T sodium salt trihydrate, 100.29%, MP Biomedicals™
Product Code. 11476512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476512 500 g 500g
11486512 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11476512 Supplier MP Biomedicals Supplier No. 0215011590
  • ONLINE EXCLUSIVE PRICE
    126.00 EUR
    119.70 EUR / 500g
    Save 6.30 EUR
    5% Off
 Restricted Item
Estimated Shipment: 14-12-2026
Chemical Identifiers
CAS 127-65-1
Molecular Formula C7H7ClNNaO2S
Molecular Weight (g/mol) 227.638
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
Synonym chloramine-t, chloramine t, chloralone, chlorasan, chlorozone, acti-chlore, chloraseptine, chlorazene, chlorazone, chlorseptol
PubChem CID 3641960
ChEBI CHEBI:53767
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]
Specifications
Color White
Density Bulk density 540 to 680kg/m3
Assay Percent Range 1.0029
Quantity 500 g
Formula Weight 281.7
Physical Form Fine Powder
Chemical Name or Material N-Chloro-p-toluenesulfonamide