Learn More
Chloracetmesidide, TRC
Specifications
Specifications
| Chemical Name or Material | Chloracetmesidide |
| CAS | 3910-51-8 |
| Molecular Formula | C11H14ClNO |
| Quantity | 250 mg |
| Synonym | 2-Chloro-N-(2,4,6-trimethylphenyl)acetamide, 2-Chloro-2',4',6'-trimethylacetanilide, N-Mesitylchloroacetamide, alpha-Chloroacetmesidide |
| InChI Formula | InChI=1S/C11H14ClNO/c1-7-4-8(2)11(9(3)5-7)13-10(14)6-12/h4-5H,6H2,1-3H3,(H,13,14) |
| SMILES | Cc1cc(C)c(NC(=O)CCl)c(C)c1 |
| IUPAC Name | 2-chloro-N-(2,4,6-trimethylphenyl)acetamide |
| Molecular Weight (g/mol) | 211.69 |
| Formula Weight | 211.0764 |