missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CHIR 98014

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
CHIR 98014 is a highly potent and selective GSK-3 inhibitor (IC50 values are 0.58 and 0.65 nM for the β and α isoforms, respectively). Exhibits >500-fold selectivity for GSK-3β over a range of other kinases. Improves insulin sensitivity of muscle strips from diabetic rats in vitro and decreases insulin and glucose levels in the oral glucose tolerance test in vivo. Activates Wnt signaling in human adipose stem cells (hASC) and promotes the production of definitive endoderm. Has been used in protocols to derive epicardial cells from hPSCs.
Specifications
Specifications
| Quantity | 5 mg |
| Molecular Formula | C20H17Cl2N9O2 |
| Formula Weight | 486.31 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction