missing translation for 'onlineSavingsMsg'
Learn More

Chicken Anti-Goat IgG Biotinylated Antibody, R&D Systems™

Product Code. 18081712
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18081712 250 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18081712 Supplier R&D Systems Supplier No. BAF019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Chicken
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects goat IgG heavy and light chains. In Western blot, less than 5% cross-reactivity with mouse IgG, rabbit IgG and human IgG is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Goat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.