Learn More
Fisherbrand™ Chest ULT Freezer Racks
Product Code. 15789529 Shop All Fisherbrand Products
Change view
Click to view available options
Holds:
12 Boxes (2 in.)
2 in. Boxes (10)
2 in. Boxes (11)
2 in. Boxes (13)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15789529 2 in. Boxes (11) Each
15709539 2 in. Boxes (13) Each
15779529 2 in. Boxes (10) Each
15799529 12 Boxes (2 in.) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15789529
  • ONLINE EXCLUSIVE PRICE
    46.25 EUR
    41.45 EUR / Pack of 1
    Save 4.80 EUR
    10% Off
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of chest ULT freezers.
  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Locking rod included
Specifications
Holds 2 in. Boxes (11)
Material Stainless Steel
For Use With (Application) Standard 2 in. Cryoboxes
For Use With (Equipment) Chest ULT Freezer
Dimensions (L x W x H) 615 x 143 x 140 mm
Product Type Vertical Rack