Learn More
Fisherbrand™ Chemical Sorbent Particulate
Product Code. 15873720 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
100 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15873720 100 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15873720
Support a Safer, More Efficient Laboratory
Use promo code 28026
View all promotions
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
Absorbs quickly, thanks to its fast wicking action. Encapsulates spills on contact.
  • Ideal for oil clean up on open water and land applications
  • Fibrous sorbent for non-aggressive spills of oil based or solvent based liquids
  • Compressed bale pack
  • Loose fibrous peat moss structure
Spillage, absorbent, spill, sorbent
Specifications
Color Brown
Capacity (Metric) 100 L
Material Large Fiber Sphagnum Peat Moss
Type Chemical Sorbent
Format Bale
For Use With (Application) Fibrous Sorbent for non-aggressive spills of Oil Based or Solvent Based Liquids.
Weight (Metric) 12.3 kg