Learn More
Fisherbrand™ Chemical Sorbent Pads, High Visibility
Product Code. 15893720 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
1.14 L
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15893720 1.14 L Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15893720
  • ONLINE EXCLUSIVE PRICE
    182.00 EUR
    123.38 EUR / Pack of 100
    Save 58.62 EUR
    32% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
Yellow colour-coded sorbent pads are highly visible for safety and easy separation of hazardous waste.
  • Yellow colour for high visibility
  • 3-ply pads
  • Perforated on centreline for dividing in half - reduces waste
  • Does not react with aggressive fluids
Spillage, absorbent, spill, sorbent
Specifications
Absorbancy Hydrophilic, Oleophilic
Color Yellow
Capacity (Metric) 1.14 L
Material Polypropylene
Type Chemical Sorbent
For Use With (Application) Abrasion-resistant, lint-resistant and chemical-resistant sorbent for oil or solvent-based liquids
Load Bearing Capacity (Metric) 5.3 kg
Packaging Quantity 100 Pads/Pk.
Thickness 3-ply
Dimensions (L x W) 500 x 400 mm
Length (Metric) 500 mm
Width (Metric) 400 mm