Learn More
LMS™ Chart Recorder Paper for Cooled Incubator
Product Code. 12522597
Change view
Click to view available options
Product Type:
Roll LMS CRP28 (Tear-off)
Roll LMS CRP29 (Re-roll)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12522597 Roll LMS CRP28 (Tear-off) Each
12813456 Roll LMS CRP29 (Re-roll) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12522597 Supplier LMS™ Supplier No. CRP28
Estimated Shipment: 03-12-2026
LMS™ Cooled Incubators come with a wide range of factory-fitted optional accessories and other extras to cater for individual requirements and even more operational flexibility.
  • Roll of chart recorder paper
  • Available in tear-off or re-roll formats
  • Accessory for use with the optional  LMS™ cooled incubator chart recorder
  • Sufficient for thirty-day recording
Specifications
Product Type Roll LMS CRP28 (Tear-off)
Description Recorder paper
For Use With (Application) Chart Recorder
For Use With (Equipment) LMS™ Chart Recorder
Quantity 1 Roll