Learn More
BRANDTECH™ BRAND™ Inductive Charging Stand for HandyStep™ touch / HandyStep™ touch S
Product Code. 16353677 Shop All Brand Products
Change view
Click to view available options
For Use With (Equipment):
HandyStep Touch and HandyStep Touch S
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
16353677 HandyStep Touch and HandyStep Touch S Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16353677 Supplier BRANDTECH™ BRAND™ Supplier No. 705220
  • ONLINE EXCLUSIVE PRICE
    297.00 EUR
    272.71 EUR / Pack of 1
    Save 24.29 EUR
    8% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
Bulk Quote Request
Clutter-free and secure charging for your HandyStep™ touch devices.
  • Keep your laboratory bench organized while securely holding the HandyStep™ touch, even with a PD-Tip™ II attached
  • Inductive charging technology
  • LED status indicator
  • Compatible with both the HandyStep™ touch and HandyStep™ touch S
  • Items supplied- Includes charging stand, USB cable (without power supply unit), and operating manual
TRUSTED_SUSTAINABILITY
Specifications
Description Power supply not included
For Use With BRAND HandyStep Touch/Touch S
Material Plastic
Type Inductive Charging Stand
For Use With (Equipment) HandyStep Touch and HandyStep Touch S