Learn More
CHAPS (3-[(3-Cholamidopropyl)-dimethylammonio]-propane- Sulfonate), Millipore™
Specifications
Specifications
| CAS | 75621-03-3 |
| Melting Point | 157°C |
| Concentration | 6 mM |
| Quantity | 10 g |
| MDL Number | MFCD00012116 |
| Synonym | CHAPS, 3-[(3-Cholamidopropyl)dimethylammonio] propane sulfonate), 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, CHAPS, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate |
| Molecular Weight (g/mol) | 614.88 |
| Chemical Name or Material | CHAPS (3-[(3-Cholamidopropyl)-Dimethylammonio]-Propane- Sulfonate) |
Safety and Handling
Product Identifier
- CHAPS (3-[(3-Cholamidopropyl)- dimethylammonio]-propane- sulphonate) for biochemistry
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate