Learn More
Tocris Bioscience™ CGP 55845 hydrochloride
GREENER_CHOICE
Product Code. 18064002
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 18064002
Brand: Tocris Bioscience™ 1248/10
Potent and selective GABAB antagonist
Potent, selective GABAB receptor antagonist (IC50 = 5 nM) that prevents agonist binding (pKi = 8.35) and inhibits GABA and glutamate release (pEC50 values are 8.08 and 7.85 respectively). Inhibits GABAB responses to baclofen (IC50 = 130 nM in an isoproterenol assay) and potentiates the hypoglycemic response to glucose in vitro.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 149184-22-5
Molecular Formula C18H23Cl3NO3P
Specifications
Quantity 10 mg
Formula Weight 438.71
Purity >98%
Molecular Weight (g/mol) 438.71
Chemical Name or Material CGP 55845 hydrochloride