missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CGI 1746
GREENER_CHOICE

Product Code. 18779876
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18779876

Brand: Tocris Bioscience™ 6965/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, reversible BTK inhibitor

CGI 1746 is a potent, reversible inhibitor of Bruton's tyrosine kinase (Btk) (IC50 = 1.9 nM). CGI 1746 decreases cell survival in prostate cancer cell lines. In a cell model, CGI 1746 blocks the production of FcγR-induced cytokines IL-1β, TNFα and IL-6 (IC50 values are 36 nM, 47 nM and 353 nM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[4,5-Dihydro-4-methyl-6-[[4-(4-morpholinylcarbonyl)phenyl]amino]-5-oxo-2-pyrazinyl]-2-methylphenyl]-4-(1,1-dimethylethyl)benzamide
CAS 910232-84-7
Quantity 50 mg
Target Brutons Tyrosine Kinase (BTK) Inhibitors
Molecular Formula C34H37N5O4
Purity 0.97
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.