Learn More
Edwards Vacuum™ CG16K Capsule Dial Gauge
Product Code. 12965274
Change view
Click to view available options
Maximum Pressure:
0 to 1040 mbar
0to 25 mbar
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Maximum Pressure Unit Size
12965274 0 to 1040 mbar Each
12995274 0to 25 mbar Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12965274 Supplier Edwards Vacuum™ Supplier No. D35610000
  • ONLINE EXCLUSIVE PRICE
    857.00 EUR
    620.30 EUR / Pack of 1
    Save 236.70 EUR
    27% Off
Estimated Shipment: 16-12-2026
Barometrically compensated design gives better repeatability than bourdon gauges, especially at low pressures
Barometrically compensated design gives better repeatability than bourdon gauges, especially at low pressures. N. B. Use an adapter plus Co-Seal and clamp if retro fitting previous model CG3 with 3/8 inch BSP fitting. For adapter, please contact Customer Service.
  • Rugged gauge with easy to read 100mm dial
  • Accuracy ±2%
  • NW16 vacuum flange Mechanical gauge for use with pumps.
Specifications
Type Capsule Dial Gauge
Maximum Pressure 0 to 1040 mbar