missing translation for 'onlineSavingsMsg'
Learn More

Cevipabulin fumarate, MedChemExpress

Product Code. 16476268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476268 10 mg 10mg
16466606 100 mg 100mg
16448857 5 mg 5mg
16422397 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16476268 Supplier Medchem Express Supplier No. HY14949C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound and inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell line.

Cevipabulin fumarate (TTI-237 fumarate) is an oral, microtubule-active, antitumor compound and inhibits the binding of [3H]NSC 49842 to tubulin, with an IC50 of 18-40 nM for cytotoxicity in human tumor cell line.

Specifications

Chemical Name or Material Cevipabulin fumarate
CAS 849550-67-0
Color White
Purity Grade Notes Research
Molecular Formula C22H22ClF5N6O5
Quantity 10 mg
Synonym TTI-237 fumarate
Solubility Information DMSO : 50 mg/mL (86.07 mM; Need ultrasonic) ∣H2O : 1.43 mg/mL (2.46 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@H](C(F)(F)F)NC1=C(C2=C(F)C=C(OCCCNC)C=C2F)C(Cl)=NC3=NC=NN13.O=C(O)/C=C/C(O)=O
Molecular Weight (g/mol) 580.89
Formula Weight 580.89
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.