missing translation for 'onlineSavingsMsg'
Learn More

Cetyl alcohol, ∽99%, MP Biomedicals™

Product Code. 11423250 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423250 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423250 Supplier MP Biomedicals Supplier No. 0210130980
  • ONLINE EXCLUSIVE PRICE
    68.35 EUR
    65.10 EUR / 100g
    Save 3.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 36653-82-4
Color White
Assay Percent Range ∽99%
Molecular Formula C16H34O
MDL Number MFCD00004760
Quantity 100 g
Synonym 1-hexadecanol, cetyl alcohol, hexadecanol, cetanol, palmityl alcohol, hexadecyl alcohol, n-cetyl alcohol, cetaffine, cetylol, cetal
InChI Key BXWNKGSJHAJOGX-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCO
IUPAC Name hexadecan-1-ol
Molecular Weight (g/mol) 242.45
PubChem CID 2682
ChEBI CHEBI:16125
Formula Weight 242.44
Physical Form Crystalline Powder
Chemical Name or Material Cetyl alcohol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.