Learn More
MedChemExpress Cetrorelix Acetate
Product Code. 16451768
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451768 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16451768 Supplier MedChemExpress Supplier No. HYP0009A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress Cetrorelix Acetate (SB-75 acetate) is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM.
Specifications
Purity 99.18%
CAS 145672-81-7
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1491.09 g/mol
Product Type Reference compound
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : 50 mg/mL (33.53 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (1.34 mM∣Need ultrasonic)
Synonym SB-75 acetate