missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cetrorelix Acetate

Product Code. 16444957
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444957 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444957 Supplier MedChemExpress Supplier No. HYP0009A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cetrorelix Acetate (SB-75 acetate) is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM.

Specifications

Purity 99.18%
CAS 145672-81-7
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1491.09 g/mol
Product Type Reference compound
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : 50 mg/mL (33.53 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (1.34 mM∣Need ultrasonic)
Synonym SB-75 acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.