Learn More
MedChemExpress Cetrorelix Acetate
Product Code. 16432609
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432609 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16432609 Supplier MedChemExpress Supplier No. HYP0009A
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
MedChemExpress Cetrorelix Acetate (SB-75 acetate) is a potent gonadotropin-releasing hormone (GnRH) receptor antagonist with an IC50 of 1.21 nM.
Specifications
Purity 99.18%
CAS 145672-81-7
Content And Storage Sealed storage, away from moisture and light∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
For Use With (Application) Neuroscience-Neuromodulation
Molecular Weight (g/mol) 1491.09 g/mol
Product Type Reference compound
Quantity 1 mg
Shipping Condition Room temperature
Solubility DMSO : 50 mg/mL (33.53 mM∣Need ultrasonic) ∣H2O : 2 mg/mL (1.34 mM∣Need ultrasonic)
Synonym SB-75 acetate
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.