Learn More
Cetirizine Ethyl Ester, TRC
Product Code. 30362933
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362933 500 mg 500mg
30350809 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30362933 Supplier Toronto Research Chemicals Supplier No. TRCC291145500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 246870-46-2
Molecular Formula C23 H29 Cl N2 O3
Molecular Weight (g/mol) 416.941
Synonym (RS)-2-[2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid ethyl ester, Ethyl Ester of Cetirizine
IUPAC Name ethyl 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetate
SMILES CCOC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C23H29ClN2O3/c1-2-29-22(27)18-28-17-16-25-12-14-26(15-13-25)23(19-6-4-3-5-7-19)20-8-10-21(24)11-9-20/h3-11,23H,2,12-18H2,1H3
Formula Weight 416.187
Percent Purity >95%
Chemical Name or Material Ethyl Ester of Cetirizine
Grade HPLC