missing translation for 'onlineSavingsMsg'
Learn More

Cetirizine dihydrochloride, 99+%, Thermo Scientific™

179.00 EUR - 512.00 EUR

Chemical Identifiers

CAS 83881-52-1
Molecular Formula C21H27Cl3N2O3
Molecular Weight (g/mol) 461.81
MDL Number MFCD00941428
InChI Key PGLIUCLTXOYQMV-UHFFFAOYNA-N
Synonym cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler
PubChem CID 55182
IUPAC Name dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
SMILES [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15415749
SDS View Documents
Thermo Scientific Alfa Aesar
J63549.06
5g
179.00 EUR
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15425749
SDS View Documents
Thermo Scientific Alfa Aesar
J63549.09
10g
251.00 EUR
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15435749
SDS View Documents
Thermo Scientific Alfa Aesar
J63549.14
25g
512.00 EUR
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

Solubility
Soluble in water, DMSO, ethanol, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Chemical Identifiers

83881-52-1
461.81
PGLIUCLTXOYQMV-UHFFFAOYNA-N
55182
[H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
C21H27Cl3N2O3
MFCD00941428
cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler
dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride

Specifications

83881-52-1
MFCD00941428
cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler
PGLIUCLTXOYQMV-UHFFFAOYNA-N
dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride
55182
≥99%
C21H27Cl3N2O3
14,2022
Soluble in water,DMSO,ethanol,and methanol.
[H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1
461.81
461.82
Cetirizine dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'rtecsNumber' : AG0980000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Cetirizine dihydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contain: Cetirizine hydrochloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.