Learn More
Cetirizine dihydrochloride, 99+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J63549.06
| Quantity | 5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCetirizine dihydrochloride is used as a Histamine H-1 receptor antagonist, antihistaminic, piperazines. Inhibits activation of eosinophils and neutrophils. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.
Solubility
Soluble in water, DMSO, ethanol, and methanol.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 83881-52-1 | |
| 461.81 | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| 55182 | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 |
| C21H27Cl3N2O3 | |
| MFCD00941428 | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride |
Specifications
| 83881-52-1 | |
| MFCD00941428 | |
| 14,2022 | |
| Soluble in water,DMSO,ethanol,and methanol. | |
| [H+].[H+].[Cl-].[Cl-].OC(=O)COCCN1CCN(CC1)C(C1=CC=CC=C1)C1=CC=C(Cl)C=C1 | |
| 461.81 | |
| 461.82 | |
| Cetirizine dihydrochloride |
| C21H27Cl3N2O3 | |
| 5g | |
| cetirizine dihydrochloride, zyrtec, reactine, cetirizine hydrochloride, 2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride, alerlisin, alercet, alergex, alertisin, cetriler | |
| PGLIUCLTXOYQMV-UHFFFAOYNA-N | |
| dihydrogen 2-(2-{4-[(4-chlorophenyl)(phenyl)methyl]piperazin-1-yl}ethoxy)acetic acid dichloride | |
| 55182 | |
| ≥99% |
Safety and Handling
- Cetirizine dihydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contain: Cetirizine hydrochloride
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.